2-(2,4,5-Trichlorophenoxy)propionic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR315333-25G
OR315333-5G
APOSOR315333-25GEA
3380
CZK
APOSOR315333-25G
APOSOR315333-5G
2-(2,4,5-Trichlorophenoxy)propionic acid
2-(2,4,5-Trichlorophenoxy)propionic acid
Vzorec:
C₉H₇Cl₃O₃ MW: 269,51 g/mol Bod tání: 175…177 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002646 CAS číslo: 93-72-1 UN: 3077 ADR: 9+EHS,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...