Tisk…

(R)-(+)-1-Phenylethanol 99%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: (R)-1-Phenylethanol, (R)-(+)-α-Methylbenzyl alcohol, (R)-(+)-sec-Phenethyl alcohol

OR315316-25G OR315316-5G OR315316-100G
APOSOR315316-25GEA 1040 CZK
APOSOR315316-25G APOSOR315316-5G APOSOR315316-100G
(R)-(+)-1-Phenylethanol 99%
(R)-(+)-1-Phenylethanol
Vzorec: C₈H₁₀O
MW: 122.17
Bod varu: 207
Storage Temperature: Ambient
MDL Number: MFCD00064263
CAS číslo: 1517-69-7
UN: 2937
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR