Tisk…
Ethyl-2-isothiocyanatobenzoate 98+%
Dodavatel: Apollo Scientific
Synonyms: o-Ethoxycarbonylphenyl isothiocyanate, 2-Ethoxycarbonylphenyl isothiocyanate
OR315248-1G OR315248-5G
APOSOR315248-1GEA 2610 CZK
APOSOR315248-1G APOSOR315248-5G
Ethyl-2-isothiocyanatobenzoate 98+%
Ethyl-2-isothiocyanatobenzoate
Vzorec: C₁₀H₉NO₂S
MW: 207.25
Storage Temperature: Ambient
MDL Number: MFCD00041063
CAS číslo: 99960-09-5