Tisk…

Methyl phenylpropiolate 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: Methyl Phenylpropiolate, Phenylpropiolic acid methyl ester

OR315233-100G OR315233-25G OR315233-5G
APOSOR315233-100GEA 14060 CZK
APOSOR315233-100G APOSOR315233-25G APOSOR315233-5G
Methyl phenylpropiolate 98%
Methyl phenylpropiolate
Vzorec: C₁₀H₈O₂
MW: 160,17 g/mol
Bod varu: 103 °C
Bod tání: 23 °C
Storage Temperature: Ambient
MDL Number: MFCD00041685
CAS číslo: 4891-38-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR