Tisk…

Ethyl phenylpropiolate

Dodavatel: Apollo Scientific

Synonyms: Phenylpropiolic acid ethyl ester, Ethyl 3-phenylprop-2-ynoate, Ethyl phenylacetylenecarboxylate

OR315228-25G
APOSOR315228-25GEA 5720 CZK
APOSOR315228-25G
Ethyl phenylpropiolate
Ethyl phenylpropiolate
Vzorec: C₁₁H₁₀O₂
MW: 174,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00009185
CAS číslo: 2216-94-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR