Tisk…

4,7-Dichloro-1,10-phenanthroline

Dodavatel: Apollo Scientific
Nebezpečí

OR315158-1G OR315158-250MG OR315158-5G
APOSOR315158-1GEA 4400 CZK
APOSOR315158-1G APOSOR315158-250MG APOSOR315158-5G
4,7-Dichloro-1,10-phenanthroline
4,7-Dichloro-1,10-phenanthroline
Vzorec: C₁₂H₆Cl₂N₂
Storage Temperature: Ambient
MDL Number: MFCD00233882
CAS číslo: 5394-23-0
UN: 2811
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR