6-Bromo-1-tetralone 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
6-Bromo-3,4-dihydro-1(2H)-naphthalenone, 6-Bromo-3,4-dihydronaphthalen-1(2H)-one
OR315157-1G
OR315157-250MG
OR315157-5G
APOSOR315157-1GEA
1210
CZK
APOSOR315157-1G
APOSOR315157-250MG
APOSOR315157-5G
6-Bromo-1-tetralone 98%
6-Bromo-1-tetralone
Vzorec:
C₁₀H₉BrO MW: 225,08 g/mol Bod tání: 44…47 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD04114378 CAS číslo: 66361-67-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...