Tisk…

2-Bromodibenzofuran 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Bromodibenzo[b,d]furan

OR315129-100G OR315129-25G OR315129-5G
APOSOR315129-100GEA 3230 CZK
APOSOR315129-100G APOSOR315129-25G APOSOR315129-5G
2-Bromodibenzofuran 98%
2-Bromodibenzofuran
Vzorec: C₁₂H₇BrO
MW: 247,09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00092338
CAS číslo: 86-76-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR