Methyl-3-bromopyrazine-2-carboxylate 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Methyl 3-Bromo-2-pyrazinecarboxylate
OR315096-1G
OR315096-5G
OR315096-10G
APOSOR315096-1GEA
1700
CZK
APOSOR315096-1G
APOSOR315096-5G
APOSOR315096-10G
Methyl-3-bromopyrazine-2-carboxylate 95%
Methyl-3-bromopyrazine-2-carboxylate
Vzorec:
C₆H₅BrN₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD08753935 CAS číslo: 51171-02-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...