Tisk…

2-(Chloromethyl)pyrimidine

Dodavatel: Apollo Scientific

OR315094-1G OR315094-5G OR315094-10G
APOSOR315094-1GEA 4100 CZK
APOSOR315094-1G APOSOR315094-5G APOSOR315094-10G
2-(Chloromethyl)pyrimidine
2-(Chloromethyl)pyrimidine
Vzorec: C₅H₅ClN₂
Storage Temperature: Ambient
MDL Number: MFCD06796203
CAS číslo: 54198-88-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR