3-Aminopicolinamide 98%
Dodavatel: Apollo Scientific
Synonyms:
3-Amino-pyridine-2-carboxylic acid amide
OR315090-1G
OR315090-10G
OR315090-5G
APOSOR315090-1GEA
1950
CZK
APOSOR315090-1G
APOSOR315090-10G
APOSOR315090-5G
3-Aminopicolinamide 98%
3-Aminopicolinamide
Vzorec:
C6H7N3O MW: 137,1400 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06658329 CAS číslo: 50608-99-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...