Tisk…

N-Methyl-D-prolinol

Dodavatel: Apollo Scientific

OR315047-100G OR315047-25G
APOSOR315047-100GEA 5590 CZK
APOSOR315047-100G APOSOR315047-25G
N-Methyl-D-prolinol
N-Methyl-D-prolinol
Vzorec: C₆H₁₃NO
MW: 115.1700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368622
CAS číslo: 99494-01-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR