5-Chloro-2-formylbenzoic acid
Dodavatel: Apollo Scientific
OR313090-1G
OR313090-5G
APOSOR313090-1GEA
2030
CZK
APOSOR313090-1G
APOSOR313090-5G
5-Chloro-2-formylbenzoic acid
5-Chloro-2-formylbenzoic acid
Vzorec:
C₈H₅ClO₃ Storage Temperature: Ambient |
MDL Number:
MFCD06739601 CAS číslo: 4506-45-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...