Tisk…

4-Chloro-3-methoxybenzaldehyde

Dodavatel: Apollo Scientific

OR313086-25G
APOSOR313086-25GEA 3540 CZK
APOSOR313086-25G
4-Chloro-3-methoxybenzaldehyde
4-Chloro-3-methoxybenzaldehyde
Vzorec: C₈H₇ClO₂
MW: 170,5900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07787490
CAS číslo: 13726-16-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR