3-Chloro-5-methylaniline 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR313080-100G
OR313080-25G
OR313080-5G
APOSOR313080-100GEA
4140
CZK
APOSOR313080-100G
APOSOR313080-25G
APOSOR313080-5G
3-Chloro-5-methylaniline 98%
3-Chloro-5-methylaniline
Vzorec:
C7H8ClN MW: 141,60 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD15527197 CAS číslo: 29027-20-1 UN: 2239 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...