5-Methoxyisoindoline
Dodavatel: Apollo Scientific
Synonyms:
5-Methoxyisoindoline hydrochloride
OR313054-1G
OR313054-250MG
APOSOR313054-1GEA
6050
CZK
APOSOR313054-1G
APOSOR313054-250MG
5-Methoxyisoindoline
5-Methoxyisoindoline
Vzorec:
C₉H₁₁NO MW: 149,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06804552 CAS číslo: 127168-88-1 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...