Tisk…

N-(2-Aminoethyl)maleimide hydrochloride

Dodavatel: Apollo Scientific
Varování

Synonyms: N-(2-Aminoethyl)maleimide HCl, N-(2-Aminoethyl)maleimide Hydrochloride, 2-Maleimidoethylamine hydrochloride

OR313050-5G
APOSOR313050-5GEA 3610 CZK
APOSOR313050-5G
N-(2-Aminoethyl)maleimide hydrochloride
N-(2-Aminoethyl)maleimide hydrochloride
Vzorec: C₆H₈N₂O₂
MW: 176,60 g/mol
Bod tání: 163 °C (rozkl.)
Storage Temperature: Refrigerator
MDL Number: MFCD09800491
CAS číslo: 134272-64-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR