Ethyl-3-methyl-1H-pyrrole-2-carboxylate 98%
Dodavatel: Apollo Scientific
Synonyms:
Ethyl 3-methyl-1H-pyrrole-2-carboxylate
OR313039-1G
OR313039-5G
OR313039-25G
APOSOR313039-1GEA
2200
CZK
APOSOR313039-1G
APOSOR313039-5G
APOSOR313039-25G
Ethyl-3-methyl-1H-pyrrole-2-carboxylate 98%
Ethyl-3-methyl-1H-pyrrole-2-carboxylate
Vzorec:
C₈H₁₁NO₂ MW: 153,1784 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01006747 CAS číslo: 3284-47-7 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...