tert-Butyl-3-formylpyrrolidine-1-carboxylate 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(+/-)-3-Formylpyrrolidine-1-carboxylic acid tert-Butyl ester, (+/-)-1-Boc-pyrrolidine-3-carboxaldehyde, (+/-)-1-(tert-Butoxycarbonyl)-3-formylpyrrolidine, (+/-)-1-Boc-3-formylpyrrolidine, tert-Butyl 3-formylpyrrolidine-1-carboxylate, (+/-)-tert-Butyl 3-Formylpyrrolidine-1-carboxylate, (+/-)-1-(tert-Butoxycarbonyl)pyrrolidine-3-carboxaldehyde
OR313035-5G
APOSOR313035-5GEA
0
CZK
APOSOR313035-5G
tert-Butyl-3-formylpyrrolidine-1-carboxylate 98%
tert-Butyl-3-formylpyrrolidine-1-carboxylate
Vzorec:
C₁₀H₁₇NO₃ MW: 199,2500 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD03791257 CAS číslo: 59379-02-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...