tert-Butyl-6-amino-3,4-dihydroisoquinoline-2(1H)-carboxylate 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
tert-Butyl 6-amino-3,4-dihydroisoquinoline-2(1H)-carboxylate, 6-Amino-2-N-BOC-1,2,3,4-tetrahydro-isoquinoline
OR313033-1G
OR313033-5G
OR313033-25G
APOSOR313033-1GEA
2530
CZK
APOSOR313033-1G
APOSOR313033-5G
APOSOR313033-25G
tert-Butyl-6-amino-3,4-dihydroisoquinoline-2(1H)-carboxylate 98%
tert-Butyl-6-amino-3,4-dihydroisoquinoline-2(1H)-carboxylate
Vzorec:
C₁₄H₂₀N₂O₂ MW: 248,3200 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03426351 CAS číslo: 164148-92-9 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...