Tisk…

Trimorpholinophosphine oxide 97%

Dodavatel: Apollo Scientific

Synonyms: Phosphoric Trimorpholide

OR3128-100G OR3128-25G
APOSOR3128-100GEA 4900 CZK
APOSOR3128-100G APOSOR3128-25G
Trimorpholinophosphine oxide 97%
Trimorpholinophosphine oxide
Vzorec: C₁₂H₂₄N₃O₄P
MW: 305.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00047406
CAS číslo: 4441-12-7

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR