3,4-Dihydro-2H-pyran-5-carbaldehyde 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3,4-Dihydro-2H-pyran-5-carboxaldehyde
OR3126-1G
OR3126-250MG
OR3126-5G
APOSOR3126-1GEA
2820
CZK
APOSOR3126-1G
APOSOR3126-250MG
APOSOR3126-5G
3,4-Dihydro-2H-pyran-5-carbaldehyde 98%
3,4-Dihydro-2H-pyran-5-carbaldehyde
Vzorec:
C₆H₈O₂ MW: 112,13 g/mol Bod varu: 90 °C/15mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00085019 CAS číslo: 25090-33-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...