Bicyclo[1.1.1]pentan-1-amine hydrochloride 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR312073-100MG
OR312073-1G
OR312073-250MG
APOSOR312073-100MGEA
622
CZK
APOSOR312073-100MG
APOSOR312073-1G
APOSOR312073-250MG
Bicyclo[1.1.1]pentan-1-amine hydrochloride 95%
Bicyclo[1.1.1]pentan-1-amine hydrochloride
Vzorec:
C₅H₉N·HCl Bod tání: 256…261 °C Storage Temperature: Ambient |
MDL Number:
MFCD19204065 CAS číslo: 22287-35-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...