Tisk…

Cyclobutanemethanamine 95%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Cyclobutylmethanamine

OR312056-1G OR312056-5G OR312056-25G
APOSOR312056-1GEA 4140 CZK
APOSOR312056-1G APOSOR312056-5G APOSOR312056-25G
Cyclobutanemethanamine 95%
Cyclobutanemethanamine
Vzorec: C₅H₁₁N
MW: 85,15 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06212616
CAS číslo: 4415-83-2
UN: 3259
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR