Cyclobutylacetic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Cyclobutylacetic acid
OR312055-1G
OR312055-5G
APOSOR312055-1GEA
2980
CZK
APOSOR312055-1G
APOSOR312055-5G
Cyclobutylacetic acid
Cyclobutylacetic acid
Vzorec:
C₆H₁₀O₂ MW: 114,1400 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD06411067 CAS číslo: 6540-33-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...