Tisk…

3-Ethynyloxetan-3-ol

Dodavatel: Apollo Scientific
Nebezpečí

OR312013-1G OR312013-250MG
APOSOR312013-1GEA 4730 CZK
APOSOR312013-1G APOSOR312013-250MG
3-Ethynyloxetan-3-ol
3-Ethynyloxetan-3-ol
Vzorec: C₅H₆O₂
Storage Temperature: Refrigerator
MDL Number: MFCD20444449
CAS číslo: 1352492-38-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR