(2E)-3-(2-Ethoxyphenyl)acrylic acid 97%
Dodavatel: Apollo Scientific
Synonyms:
2-Ethoxycinnamic acid
OR311101-5G
OR311101-1G
APOSOR311101-5GEA
2240
CZK
APOSOR311101-5G
APOSOR311101-1G
(2E)-3-(2-Ethoxyphenyl)acrylic acid 97%
(2E)-3-(2-Ethoxyphenyl)acrylic acid
Vzorec:
C₁₁H₁₂O₃ MW: 192,22 g/mol Bod tání: 134…136 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016839 CAS číslo: 69038-81-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...