Tisk…

(4-Methylpiperazin-1-yl)(4-nitrophenyl)methanone

Dodavatel: Apollo Scientific

OR310338-1G OR310338-5G
APOSOR310338-1GEA 1990 CZK
APOSOR310338-1G APOSOR310338-5G
(4-Methylpiperazin-1-yl)(4-nitrophenyl)methanone
(4-Methylpiperazin-1-yl)(4-nitrophenyl)methanone
Vzorec: C₁₂H₁₅N₃O₃
Storage Temperature: Ambient
MDL Number: MFCD00585638
CAS číslo: 21091-98-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR