6-Chloro-2-(dimethylamino)-1,8-naphthyridine-3-carbonitrile
Dodavatel: Apollo Scientific
OR310226-500MG
OR310226-1G
APOSOR310226-500MGEA
18820
CZK
APOSOR310226-500MG
APOSOR310226-1G
6-Chloro-2-(dimethylamino)-1,8-naphthyridine-3-carbonitrile
6-Chloro-2-(dimethylamino)-1,8-naphthyridine-3-carbonitrile
Vzorec:
C₁₁H₉ClN₄ Bod tání: 218…220 °C Storage Temperature: Ambient |
MDL Number:
MFCD20232926 CAS číslo: 1335220-61-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...