3-Bromo-1-tert-butyl-1H-pyrazolo[3,4-d]pyrimidin-4-amine 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Amino-3-bromo-1-(tert-butyl)-1H-pyrazolo[3,4-d]pyrimidine
OR310170-500MG
OR310170-1G
OR310170-100MG
APOSOR310170-500MGEA
7340
CZK
APOSOR310170-500MG
APOSOR310170-1G
APOSOR310170-100MG
3-Bromo-1-tert-butyl-1H-pyrazolo[3,4-d]pyrimidin-4-amine 95%
3-Bromo-1-tert-butyl-1H-pyrazolo[3,4-d]pyrimidin-4-amine
Vzorec:
C₉H₁₂BrN₅ MW: 270,14 g/mol Bod tání: 180…182 °C Storage Temperature: Ambient |
MDL Number:
MFCD20921583 CAS číslo: 862728-61-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...