2,5-Dichlorothiophene-3-boronic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(2,5-Dichlorothiophen-3-yl)boronic acid
OR310159-1G
OR310159-250MG
OR310159-5G
APOSOR310159-1GEA
6020
CZK
APOSOR310159-1G
APOSOR310159-250MG
APOSOR310159-5G
2,5-Dichlorothiophene-3-boronic acid
2,5-Dichlorothiophene-3-boronic acid
Vzorec:
C₄H₃BCl₂O₂S Storage Temperature: Refrigerator |
MDL Number:
MFCD09837622 CAS číslo: 177735-28-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...