Tisk…

3-Methylquinoxaline-2-carboxylic acid 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Methyl-2-quinoxalinecarboxylic acid

OR310133-1G OR310133-250MG
APOSOR310133-1GEA 4630 CZK
APOSOR310133-1G APOSOR310133-250MG
3-Methylquinoxaline-2-carboxylic acid 95%
3-Methylquinoxaline-2-carboxylic acid
Vzorec: C₁₀H₈N₂O₂
MW: 188,19 g/mol
Bod tání: 155 °C (rozkl.)
Storage Temperature: Freezer
MDL Number: MFCD03840415
CAS číslo: 74003-63-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR