Tisk…
2-(10H-Phenothiazin-10-yl)ethan-1-amine hydrochloride
Dodavatel: Apollo Scientific
OR310012-1G OR310012-500MG OR310012-5G
APOSOR310012-1GEA 9560 CZK
APOSOR310012-1G APOSOR310012-500MG APOSOR310012-5G
2-(10H-Phenothiazin-10-yl)ethan-1-amine hydrochloride
2-(10H-Phenothiazin-10-yl)ethan-1-amine hydrochloride
Vzorec: C₁₄H₁₅ClN₂S
MW: 278.81 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22192242
CAS číslo: 50971-79-4