Ethyl-6-{2,4-dioxo-3-azatricyclo[7.3.1.0
Dodavatel: Apollo Scientific
Synonyms:
Ethyl 6-(1,3-dioxo-1H-benzo[de]isoquinolin-2(3H)-yl)hexanoate
OR310011-1G
OR310011-500MG
APOSOR310011-1GEA
9560
CZK
APOSOR310011-1G
APOSOR310011-500MG
Ethyl-6-{2,4-dioxo-3-azatricyclo[7.3.1.0
Ethyl-6-{2,4-dioxo-3-azatricyclo[7.3.1.0^{5,13}]trideca-1(13),5,7,9,11- pentaen-3-yl}hexanoate
Vzorec:
C₂₀H₂₁NO₄ MW: 339,39 g/mol Bod tání: 88…90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00376486 CAS číslo: 300815-53-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...