4,6-Dihydroxy-5-methoxypyrimidine
Dodavatel: Apollo Scientific
OR3100-1G
OR3100-250MG
OR3100-500MG
OR3100-2.5G
OR3100-5G
APOSOR3100-1GEA
4860
CZK
APOSOR3100-1G
APOSOR3100-250MG
APOSOR3100-500MG
APOSOR3100-2.5G
APOSOR3100-5G
4,6-Dihydroxy-5-methoxypyrimidine
4,6-Dihydroxy-5-methoxypyrimidine
Vzorec:
C₅H₆N₂O₃ MW: 142,12 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07368271 CAS číslo: 5193-84-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...