Tisk…

5-(5-Chloro-2-thienyl)-1H-pyrazole

Dodavatel: Apollo Scientific
Varování

OR31-250MG OR31-1G
APOSOR31-250MGEA 1450 CZK
APOSOR31-250MG APOSOR31-1G
5-(5-Chloro-2-thienyl)-1H-pyrazole
5-(5-Chloro-2-thienyl)-1H-pyrazole
Vzorec: C₇H₅ClN₂S
MW: 184,65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03791215
CAS číslo: 166196-61-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR