Tisk…

tert-Butyl-3-amino-3-ethylpiperidine-1-carboxylate

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Amino-3-ethylpiperidine, N1-BOC protected

OR30969-1G OR30969-250MG OR30969-500MG OR30969-2.5G OR30969-5G
APOSOR30969-1GEA 12300 CZK
APOSOR30969-1G APOSOR30969-250MG APOSOR30969-500MG APOSOR30969-2.5G APOSOR30969-5G
tert-Butyl-3-amino-3-ethylpiperidine-1-carboxylate
tert-Butyl-3-amino-3-ethylpiperidine-1-carboxylate
Vzorec: C₁₂H₂₄N₂O₂
MW: 228.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11840263
CAS číslo: 1158759-09-1

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR