Tisk…

4,7-Dichloro-8-methylquinoline

Dodavatel: Apollo Scientific

OR309455-1G OR309455-5G
APOSOR309455-1GEA 2570 CZK
APOSOR309455-1G APOSOR309455-5G
4,7-Dichloro-8-methylquinoline
4,7-Dichloro-8-methylquinoline
Vzorec: C₁₀H₇Cl₂N
MW: 212,800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02684150
CAS číslo: 643039-79-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR