Tisk…

7-Methylcoumarin 98%

Dodavatel: Apollo Scientific

Synonyms: 7-Methylcumarin, 7-Methyl-2H-chromen-2-one

OR30939-25G
APOSOR30939-25GEA 5230 CZK
APOSOR30939-25G
7-Methylcoumarin 98%
7-Methylcoumarin
Vzorec: C₁₀H₈O₂
MW: 160,17 g/mol
Bod tání: 128…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00006880
CAS číslo: 2445-83-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR