3-Amino-3-(2-chlorophenyl)propionic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-Amino-3-(2-chlorophenyl)propanoic acid
OR309360-1G
OR309360-5G
APOSOR309360-1GEA
1660
CZK
APOSOR309360-1G
APOSOR309360-5G
3-Amino-3-(2-chlorophenyl)propionic acid
3-Amino-3-(2-chlorophenyl)propionic acid
Vzorec:
C₉H₁₀ClNO₂ MW: 199,64 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01871299 CAS číslo: 68208-20-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...