Tisk…

Ethyl-6-bromo-4-chloroquinoline-3-carboxylate

Dodavatel: Apollo Scientific

Synonyms: 6-Bromo-4-chloroquinoline-3-carboxylic acid ethyl ester

OR309351-5G
APOSOR309351-5GEA 4600 CZK
APOSOR309351-5G
Ethyl-6-bromo-4-chloroquinoline-3-carboxylate
Ethyl-6-bromo-4-chloroquinoline-3-carboxylate
Vzorec: C₁₂H₉BrClNO₂
MW: 314,563 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00173367
CAS číslo: 206257-39-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR