Tisk…

Ethyl-4-chloroquinoline-3-carboxylate

Dodavatel: Apollo Scientific

Synonyms: 4-Chloroquinoline-3-carboxylic acid ethyl ester, Ethyl 4-chloroquinoline-3-carboxylate

OR309348-5G
APOSOR309348-5GEA 2280 CZK
APOSOR309348-5G
Ethyl-4-chloroquinoline-3-carboxylate
Ethyl-4-chloroquinoline-3-carboxylate
Vzorec: C₁₂H₁₀ClNO₂
MW: 235,67 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00173407
CAS číslo: 13720-94-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR