Tisk…

2-Chloro-3-ethyl-quinoline

Dodavatel: Apollo Scientific

Synonyms: 2-Chloro-3-ethylquinoline

OR309257-5G
APOSOR309257-5GEA 4900 CZK
APOSOR309257-5G
2-Chloro-3-ethyl-quinoline
2-Chloro-3-ethyl-quinoline
Vzorec: C₁₁H₁₀ClN
MW: 191,66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02684198
CAS číslo: 67525-28-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR