Tisk…

6-Bromo-2-chloro-3-methylquinoline

Dodavatel: Apollo Scientific
Varování

OR309201-1G OR309201-5G
APOSOR309201-1GEA 8240 CZK
APOSOR309201-1G APOSOR309201-5G
6-Bromo-2-chloro-3-methylquinoline
6-Bromo-2-chloro-3-methylquinoline
Vzorec: C₁₀H₇BrClN
MW: 256.5300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02684154
CAS číslo: 113092-96-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR