(S)-Pyrrolidin-3-ylmethanol
Dodavatel: Apollo Scientific
|
Varování
|
OR309159-1G
OR309159-5G
APOSOR309159-1GEA
3150
CZK
APOSOR309159-1G
APOSOR309159-5G
(S)-Pyrrolidin-3-ylmethanol
(S)-Pyrrolidin-3-ylmethanol
Vzorec:
C₅H₁₁NO MW: 101,15 g/mol Bod varu: 165…171 °C Hustota: 0,979 g/cm³ (25 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD09260722 CAS číslo: 110013-19-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...