Tisk…

tert-Butyl-3-(hydroxymethyl)pyrrolidine-1-carboxylate 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: tert-Butyl 3-(hydroxymethyl)pyrrolidine-1-carboxylate, 1-BOC-3-Pyrrolidinemethanol, 1-Boc-3-(hydroxymethyl)pyrrolidine

OR309157-5G OR309157-1G
APOSOR309157-5GEA 1160 CZK
APOSOR309157-5G APOSOR309157-1G
tert-Butyl-3-(hydroxymethyl)pyrrolidine-1-carboxylate 97%
tert-Butyl-3-(hydroxymethyl)pyrrolidine-1-carboxylate
Vzorec: C₁₀H₁₉NO₃
MW: 201,27 g/mol
Bod varu: 285…291 °C
Storage Temperature: Ambient
MDL Number: MFCD02179040
CAS číslo: 114214-69-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR