4-Methylpiperazin-2-one 97%
Dodavatel: Apollo Scientific
Synonyms:
1-Methyl-3-oxopiperazine
OR309137-1G
OR309137-10G
OR309137-5G
APOSOR309137-1GEA
870
CZK
APOSOR309137-1G
APOSOR309137-10G
APOSOR309137-5G
4-Methylpiperazin-2-one 97%
4-Methylpiperazin-2-one
Vzorec:
C₅H₁₀N₂O MW: 114,15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11870124 CAS číslo: 34770-60-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...