Tisk…

Ethyl 2-(oxetan-3-ylidene)acetate 97%

Dodavatel: Apollo Scientific

Synonyms: Ethyl 2-(oxetan-3-ylidene)acetate

OR309117-25G OR309117-5G
APOSOR309117-25GEA 7670 CZK
APOSOR309117-25G APOSOR309117-5G
Ethyl 2-(oxetan-3-ylidene)acetate 97%
Ethyl-2-(oxetan-3-ylidene)acetate
Vzorec: C₇H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD12755197
CAS číslo: 922500-91-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR