Tisk…

2-(Oxetan-3-yl)ethanol 97%

Dodavatel: Apollo Scientific
Varování

OR309064-1G OR309064-250MG
APOSOR309064-1GEA 16340 CZK
APOSOR309064-1G APOSOR309064-250MG
2-(Oxetan-3-yl)ethanol 97%
2-(Oxetan-3-yl)ethanol
Vzorec: C₅H₁₀O₂
MW: 102,13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17215859
CAS číslo: 251922-46-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR