Tisk…

1-(tert-Butoxycarbonyl)azetidine-2-carboxylic acid 95%

Dodavatel: Apollo Scientific

Synonyms: 1-[(tert-butoxy)carbonyl]azetidine-2-carboxylic acid

OR309044-1G OR309044-5G
APOSOR309044-1GEA 1820 CZK
APOSOR309044-1G APOSOR309044-5G
1-(tert-Butoxycarbonyl)azetidine-2-carboxylic acid 95%
1-(tert-Butoxycarbonyl)azetidine-2-carboxylic acid
Vzorec: C₉H₁₅NO₄
MW: 201,2197 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01861756
CAS číslo: 159749-28-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR